(4-Amino-1H-pyrazol-1-yl)acetic acid hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 2-(4-Amino-1H-pyrazol-1-yl)acetic acid hydrochloride

A496492-100MG A496492-250MG A496492-5G A496492-1G
77241-468EA 14.42 USD
77241-468 77241-470 77241-472 77747-016
(4-Amino-1H-pyrazol-1-yl)acetic acid hydrochloride 98%
(4-Amino-1H-pyrazol-1-yl)acetic acid hydrochloride

Precaución: for Research Use Only

Formula: C₅H₈ClN₃O₂
MW: 177.59 g/mol
MDL Number: MFCD22421967
CAS Number: 1417569-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR