BMS-5 98%

Supplier: Ambeed
Warning

A496357-100MG A496357-1MG A496357-50MG A496357-5MG
77241-460EA 299 USD
77241-460 77241-462 77241-464 77241-466
BMS-5 98%
BMS-5

Precaución: for Research Use Only

Formula: C₁₇H₁₄Cl₂F₂N₄OS
MDL Number: MFCD17019327
CAS Number: 1338247-35-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR