4,6-Dichloro-2-(methylthio)pyrimidin-5-amine 97%

Supplier: Ambeed
Warning

A496329-100MG A496329-1G A496329-250MG
77241-454EA 56.65 USD
77241-454 77241-456 77241-458
4,6-Dichloro-2-(methylthio)pyrimidin-5-amine 97%
4,6-Dichloro-2-(methylthio)pyrimidin-5-amine

Precaución: for Research Use Only

Formula: C₅H₅Cl₂N₃S
MW: 210.08 g/mol
MDL Number: MFCD24646377
CAS Number: 333388-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR