2-Methyl-3-nitroanisole 98%

Supplier: Ambeed
Warning

Synonyms: 1-Methoxy-2-methyl-3-nitrobenzene, Methyl 3-nitro-o-tolyl ether, 2-Methoxy-6-nitrotoluene

A496303-100G A496303-1G A496303-25G A496303-5G
77253-228EA 128 USD
77253-228 77253-230 77253-232 77617-124
2-Methyl-3-nitroanisole 98%
2-Methyl-3-nitroanisole

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MDL Number: MFCD00007159
CAS Number: 4837-88-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR