IRAK-1/4 Inhibitor 98%

Supplier: Ambeed
Warning

A496272-100MG A496272-10MG A496272-1MG A496272-50MG A496272-5MG
77241-444EA 151 USD
77241-444 77241-446 77241-448 77241-450 77241-452
IRAK-1/4 Inhibitor 98%
IRAK-1/4 Inhibitor

Precaución: for Research Use Only

Formula: C₂₀H₂₁N₅O
MDL Number: MFCD09752602
CAS Number: 509093-47-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR