2,4,6-Trimethylphenylhydrazine hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: Mesitylhydrazine xhydrochloride

A496228-100G A496228-10G A496228-1G A496228-250MG A496228-25G A496228-5G
77253-216EA 801.34 USD
77253-216 77253-218 77253-220 77253-222 77253-224 77253-226
2,4,6-Trimethylphenylhydrazine hydrochloride 95%
2,4,6-Trimethylphenylhydrazine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₅ClN₂
MW: 186.69 g/mol
MDL Number: MFCD00052268
CAS Number: 76195-82-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR