3-Butoxyphenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-Butoxybenzeneboronic Acid

A495982-10G A495982-1G A495982-25G A495982-5G
77279-658EA 57.68 USD
77279-658 77279-660 77279-662 77279-664
3-Butoxyphenylboronic acid 97%
3-Butoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₅BO₃
MW: 194.04 g/mol
MDL Number: MFCD03427053
CAS Number: 352534-81-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR