5-Iodo-2-methoxybenzaldehyde 95%

Supplier: Ambeed
Danger

A495715-100MG A495715-1G A495715-250MG A495715-5G
77241-436EA 9.27 USD
77241-436 77241-438 77241-440 77241-442
5-Iodo-2-methoxybenzaldehyde 95%
5-Iodo-2-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇IO₂
MDL Number: MFCD04128999
CAS Number: 42298-41-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR