4'-Bromobenzo-15-crown-5 (4'-bromobenzo-1,4,7,10,13-pentaoxacyclopentadecane) 98%

Supplier: Ambeed
Warning

Synonyms: 4'-Bromobenzo-15-crown 5-Ether, 2,3-(4-Bromobenzo)-1,4,7,10,13-pentaoxacyclopentadec-2-ene, 15-Bromo-2,3,5,6,8,9,11,12-octahydrobenzo[b][1,4,7,10,13]pentaoxacyclopentadecine

A495575-100MG A495575-250MG
77292-088EA 11 USD
77292-088 77292-090
4'-Bromobenzo-15-crown-5 (4'-bromobenzo-1,4,7,10,13-pentaoxacyclopentadecane) 98%
4'-Bromobenzo-15-crown-5 (4'-bromobenzo-1,4,7,10,13-pentaoxacyclopentadecane)

Precaución: for Research Use Only

Formula: C₁₄H₁₉BrO₅
MW: 347.2000 g/mol
MDL Number: MFCD00143270
CAS Number: 60835-72-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR