(S)-[2,3':1',1'':3'',2'''-Quaternaphthalene]-2',2''-diol ≥98%, ee 99%

Supplier: Ambeed
Warning

A495442-1G A495442-250MG
77253-212EA 412 USD
77253-212 77253-214
(S)-[2,3':1',1'':3'',2'''-Quaternaphthalene]-2',2''-diol ≥98%, ee 99%
(S)-[2,3':1',1'':3'',2'''-Quaternaphthalene]-2',2''-diol

Precaución: for Research Use Only

Formula: C₄₀H₂₆O₂
MW: 538.63 g/mol
CAS Number: 863659-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR