2,4-Dichloro-α-methylbenzyl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: 2,4-Dichlorophenyl methyl carbinol, 2,4-Dichloro-alpha-methylbenzyl alcohol, 1-(2,4-Dichlorophenyl)ethyl alcohol, 1-(2,4-Dichlorophenyl)ethanol

A494334-10G A494334-1G A494334-5G
77253-200EA 64.89 USD
77253-200 77253-202 77253-204
2,4-Dichloro-α-methylbenzyl alcohol 98%
2,4-Dichloro-α-methylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₈Cl₂O
MW: 191.06 g/mol
MDL Number: MFCD00004511
CAS Number: 1475-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR