4-(Trifluoromethyl)phenylacetylene 97%

Supplier: Ambeed
Danger

Synonyms: 4-Ethynylbenzotrifluoride, 4-Ethynyl-alpha,alpha,alpha-trifluorotoluene, 1-ethynyl-4-(trifluoromethyl)benzene

A494276-10G A494276-1G A494276-250MG A494276-25G A494276-5G
77265-614EA 91.67 USD
77265-614 77265-616 77265-618 77265-620 77265-622
4-(Trifluoromethyl)phenylacetylene 97%
4-(Trifluoromethyl)phenylacetylene

Precaución: for Research Use Only

Formula: C₉H₅F₃
MW: 170.134 g/mol
MDL Number: MFCD01861903
CAS Number: 705-31-7
UN: 1993
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR