Methyl-3,5-dichlorobenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3,5-dichlorobenzoate

A494237-1G A494237-25G A494237-5G A494237-100G
77279-632EA 5.15 USD
77279-632 77279-634 77279-636 77749-009
Methyl-3,5-dichlorobenzoate 98%
Methyl-3,5-dichlorobenzoate

Precaución: for Research Use Only

Formula: C₈H₆Cl₂O₂
MW: 205.04 g/mol
MDL Number: MFCD00032710
CAS Number: 2905-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR