2,6-Dimethyl-5-heptenal 85%

Supplier: Ambeed
Warning

Synonyms: 2,6-Dimethylhept-5-enal

A494107-100G A494107-25G A494107-500G
77253-196EA 38.11 USD
77253-196 77253-198 77747-012
2,6-Dimethyl-5-heptenal 85%
2,6-Dimethyl-5-heptenal

Precaución: for Research Use Only

Formula: C₉H₁₆O
MW: 140.23 g/mol
MDL Number: MFCD00006981
CAS Number: 106-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR