4-(4-Cyanophenyl)morpholine 97%

Supplier: Ambeed
Danger

A494033-10G A494033-25G
77292-072EA 59 USD
77292-072 77292-074
4-(4-Cyanophenyl)morpholine 97%
4-(4-Cyanophenyl)morpholine

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂O
MDL Number: MFCD00100115
CAS Number: 10282-31-2
UN: 3439
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR