5,6-Dichloro-3,4-dihydro-2H-benzo[e][1,2,4]thiadiazine-7-sulfonamide 1,1-dioxide 97%

Supplier: Ambeed
Warning

A493978-1G A493978-250MG
77253-192EA 805 USD
77253-192 77253-194
5,6-Dichloro-3,4-dihydro-2H-benzo[e][1,2,4]thiadiazine-7-sulfonamide 1,1-dioxide 97%
5,6-Dichloro-3,4-dihydro-2H-benzo[e][1,2,4]thiadiazine-7-sulfonamide 1,1-dioxide

Precaución: for Research Use Only

Formula: C₇H₇Cl₂N₃O₄S₂
MDL Number: MFCD18379277
CAS Number: 5233-42-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR