3,5-Dichlorophenethyl alcohol 95+%

Supplier: Ambeed
Warning

Synonyms: 2-(3,5-Dichlorophenyl)ethanol

A493378-10G A493378-1G A493378-5G
77279-626EA 93 USD
77279-626 77279-628 77279-630
3,5-Dichlorophenethyl alcohol 95+%
3,5-Dichlorophenethyl alcohol

Precaución: for Research Use Only

Formula: C₈H₈Cl₂O
MW: 191.055 g/mol
MDL Number: MFCD06201039
CAS Number: 93427-13-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR