Bis(4-chlorophenyl) sulfone 98%

Supplier: Ambeed
Warning

Synonyms: 4,4'-Dichloro diphenyl sulphone, 4-Chlorophenyl sulphone, 4,4'-sulphonylbis(chlorobenzene)

A493180-1000G A493180-100G A493180-25G A493180-500G
77292-058EA 51.5 USD
77292-058 77292-060 77292-062 77292-064
Bis(4-chlorophenyl) sulfone 98%
Bis(4-chlorophenyl)sulphone

Precaución: for Research Use Only

Formula: C₁₂H₈Cl₂O₂S
MW: 287.17 g/mol
MDL Number: MFCD00000619
CAS Number: 80-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR