3-Methyl-2H-chromen-2-one 97%

Supplier: Ambeed
Danger

A493170-5G A493170-1G
77279-624EA 101 USD
77279-624 77617-118
3-Methyl-2H-chromen-2-one 97%
3-Methyl-2H-chromen-2-one

Precaución: for Research Use Only

Formula: C₁₀H₈O₂
MW: 160.16 g/mol
MDL Number: MFCD00059758
CAS Number: 2445-82-1
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR