N,N-Dimethyl-2,2,2-trifluoroacetamide 98%

Supplier: Ambeed
Danger

Synonyms: 2,2,2-Trifluoro-N,N-dimethylacetamide

A492965-1G A492965-25G A492965-5G
77292-052EA 5.15 USD
77292-052 77292-054 77292-056
N,N-Dimethyl-2,2,2-trifluoroacetamide 98%
N,N-Dimethyl-2,2,2-trifluoroacetamide

Precaución: for Research Use Only

Formula: C₄H₆F₃NO
MW: 141.0900 g/mol
MDL Number: MFCD00043555
CAS Number: 1547-87-1
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR