1-Methyl-4-(4-nitrophenyl)piperazine 98%

Supplier: Ambeed
Warning

A492959-100G A492959-25G A492959-5G
77241-424EA 97 USD
77241-424 77241-426 77241-428
1-Methyl-4-(4-nitrophenyl)piperazine 98%
1-Methyl-4-(4-nitrophenyl)piperazine

Precaución: for Research Use Only

Formula: C₁₁H₁₅N₃O₂
MDL Number: MFCD00101003
CAS Number: 16155-03-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR