2-Fluoro-4-(trifluoromethyl)benzyl bromide 95%

Supplier: Ambeed
Danger

A492911-10G A492911-5G A492911-1G
77265-610EA 106 USD
77265-610 77265-612 77749-006
2-Fluoro-4-(trifluoromethyl)benzyl bromide 95%
2-Fluoro-4-(trifluoromethyl)benzyl bromide

Precaución: for Research Use Only

Formula: C₈H₅BrF₄
MDL Number: MFCD00061173
CAS Number: 239087-07-1
UN: 1760
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR