1-(2-Naphthyl)ethanamine 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-(Naphthalen-2-yl)ethanamine
A492529-100MG
A492529-1G
A492529-250MG
77241-418EA
16.48
USD
77241-418
77241-420
77747-010
1-(2-Naphthyl)ethanamine 95%
1-(2-Naphthyl)ethanamine
Precaución: for Research Use Only
Formula:
C₁₂H₁₃N MW: 171.24 g/mol |
MDL Number:
MFCD00014321 CAS Number: 1201-74-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...