1-((9H-Fluoren-9-yl)methyl)piperidine 97%

Supplier: Ambeed
Warning

A492493-250MG A492493-100MG A492493-5G
77279-618EA 120.51 USD
77279-618 77594-544 77749-005
1-((9H-Fluoren-9-yl)methyl)piperidine 97%
1-((9H-Fluoren-9-yl)methyl)piperidine

Precaución: for Research Use Only

Formula: C₁₉H₂₁N
MW: 263.37 g/mol
MDL Number: MFCD22587875
CAS Number: 35661-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR