4'-Chloro-2'-hydroxyacetophenone 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-(4-Chloro-2-hydroxyphenyl)ethan-1-one, 4-Chloro-2-hydroxyacetophenone
A492473-100G
A492473-10G
A492473-1G
A492473-25G
A492473-5G
77241-408EA
482.04
USD
77241-408
77241-410
77241-412
77241-414
77241-416
4'-Chloro-2'-hydroxyacetophenone 98%
4'-Chloro-2'-hydroxyacetophenone
Precaución: for Research Use Only
Formula:
C₈H₇ClO₂ MW: 170.6 g/mol |
MDL Number:
MFCD00238557 CAS Number: 6921-66-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...