4-Phenylbutyl 4-methylbenzenesulfonate 97%
Supplier: Ambeed
|
Warning
|
A491911-1G
A491911-250MG
A491911-5G
77253-182EA
123.6
USD
77253-182
77253-184
77253-186
4-Phenylbutyl 4-methylbenzenesulfonate 97%
4-Phenylbutyl 4-methylbenzenesulfonate
Precaución: for Research Use Only
Formula:
C₁₇H₂₀O₃S MW: 304.40 g/mol |
MDL Number:
MFCD00159238 CAS Number: 112775-09-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...