5'-(4-Carboxy-3-hydroxyphenyl)-3,3''-dihydroxy-[1,1':3',1''-terphenyl]-4,4''-dicarboxylic acid 97%

Supplier: Ambeed
Warning

A491600-100MG A491600-1G A491600-250MG
77265-588EA 36 USD
77265-588 77265-590 77265-592
5'-(4-Carboxy-3-hydroxyphenyl)-3,3''-dihydroxy-[1,1':3',1''-terphenyl]-4,4''-dicarboxylic acid 97%
5'-(4-Carboxy-3-hydroxyphenyl)-3,3''-dihydroxy-[1,1':3',1''-terphenyl]-4,4''-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₂₇H₁₈O₉
MDL Number: MFCD34179860
CAS Number: 1397264-18-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR