(R)-1-(3-Chloro-4-methylphenyl)ethanamine hydrochloride 95%

Supplier: Ambeed
Warning

A491592-100MG A491592-1G A491592-250MG A491592-5G
77279-608EA 39 USD
77279-608 77279-610 77279-612 77279-614
(R)-1-(3-Chloro-4-methylphenyl)ethanamine hydrochloride 95%
(R)-1-(3-Chloro-4-methylphenyl)ethanamine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₃Cl₂N
MDL Number: MFCD16293365
CAS Number: 856562-92-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR