4-Bromo-2-chlorothiophene 95%

Supplier: Ambeed
Warning

A490631-100MG A490631-250MG
77292-046EA 105.06 USD
77292-046 77292-048
4-Bromo-2-chlorothiophene 95%
4-Bromo-2-chlorothiophene

Precaución: for Research Use Only

Formula: C₄H₂BrClS
MW: 197.48 g/mol
MDL Number: MFCD00043884
CAS Number: 32431-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR