2-Bromoethyl methacrylate 95% stabilized
Supplier: Ambeed
|
Warning
|
Synonyms:
Methacrylic acid 2-Bromoethyl ester
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
A490357-1G
A490357-25G
A490357-5G
77265-580EA
23.69
USD
77265-580
77265-582
77265-584
2-Bromoethyl methacrylate 95% stabilized
2-Bromoethyl methacrylate
Precaución: for Research Use Only
Formula:
C₆H₉BrO₂ MW: 193.04 g/mol |
MDL Number:
MFCD00045051 CAS Number: 4513-56-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...