Methyl 2-(methylthio)-4-(trifluoromethyl)benzoate 96%

Supplier: Ambeed
Warning

A489460-1G A489460-250MG A489460-5G
77241-396EA 101.97 USD
77241-396 77241-398 77241-400
Methyl 2-(methylthio)-4-(trifluoromethyl)benzoate 96%
Methyl 2-(methylthio)-4-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₉F₃O₂S
MW: 250.24 g/mol
MDL Number: MFCD14635484
CAS Number: 192805-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR