2-(Benzhydrylthio)acetamide 98%

Supplier: Ambeed
Danger

Synonyms: 2-[(Diphenylmethyl)thio]acetamide, 2-[(Diphenylmethyl)sulphanyl]acetamide, 2-(Benzhydrylsulphanyl)acetamide

A489017-100G A489017-10G A489017-25G A489017-5G
77279-582EA 259.56 USD
77279-582 77279-584 77279-586 77279-588
2-(Benzhydrylthio)acetamide 98%
2-(Benzhydrylthio)acetamide

Precaución: for Research Use Only

Formula: C₁₅H₁₅NOS
MW: 257.351 g/mol
MDL Number: MFCD00876107
CAS Number: 68524-30-1
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR