3-tert-Butylphenol 98%

Supplier: Ambeed
Danger

Synonyms: m-tert-Butylphenol

A487314-100G A487314-5G A487314-25G
77292-034EA 40.17 USD
77292-034 77292-036 77551-474
3-tert-Butylphenol 98%
3-tert-Butylphenol

Precaución: for Research Use Only

Formula: C₁₀H₁₄O
MW: 150.22 g/mol
MDL Number: MFCD00002300
CAS Number: 585-34-2
UN: 2430
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR