(2R,3R,4R,5R,6R)-5-Acetamido-2-(acetoxymethyl)-6-((6-(((benzyloxy)carbonyl)amino)hexyl)oxy)tetrahydro-2H-pyran-3,4-diyl diacetate 97%
Supplier: Ambeed
|
Warning
|
A487291-5G
A487291-1G
77253-166EA
76
USD
77253-166
77747-003
(2R,3R,4R,5R,6R)-5-Acetamido-2-(acetoxymethyl)-6-((6-(((benzyloxy)carbonyl)amino)hexyl)oxy)tetrahydro-2H-pyran-3,4-diyl diacetate 97%
(2R,3R,4R,5R,6R)-5-Acetamido-2-(acetoxymethyl)-6-((6-(((benzyloxy)carbonyl)amino)hexyl)oxy)tetrahydro-2H-pyran-3,4-diyl diacetate
Precaución: for Research Use Only
Formula:
C₂₈H₄₀N₂O₁₁ |
MDL Number:
MFCD30489047 CAS Number: 159173-77-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...