(+)-Benzotetramisole ≥98%, ee 99%

Supplier: Ambeed
Warning

Synonyms: (R)-2-Phenyl-2,3-dihydrobenzo[d]imidazo[2,1-b]thiazole, (+)-BTM, (2R)-2,3-Dihydro-2-phenylimidazo[2,1-b]benzothiazole

A487107-100MG A487107-1G A487107-250MG A487107-5G
77253-158EA 19.57 USD
77253-158 77253-160 77253-162 77617-112
(+)-Benzotetramisole ≥98%, ee 99%
(+)-Benzotetramisole

Precaución: for Research Use Only

Formula: C₁₅H₁₂N₂S
MW: 252.34 g/mol
MDL Number: MFCD11112169
CAS Number: 885051-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR