Ethyl 2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy)acetate 97%
Supplier: Ambeed
|
Warning
|
A486917-1G
A486917-250MG
A486917-5G
77292-020EA
103
USD
77292-020
77292-022
77292-024
Ethyl 2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy)acetate 97%
Ethyl 2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy)acetate
Precaución: for Research Use Only
Formula:
C₁₆H₂₃BO₅ MW: 306.16 g/mol |
MDL Number:
MFCD06657886 CAS Number: 269410-28-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...