2-(1-Piperazinylcarbonyl)-1,4-benzodioxane hydrochloride 98%

Supplier: Ambeed
Danger

Synonyms: 1-[(1,4-Benzodioxan-2-yl)carbonyl]piperazine Hydrochloride, 2-(1-Piperazinylcarbonyl)-1,4-benzodioxane Hydrochloride

A486795-1G A486795-25G A486795-5G
77279-562EA 25 USD
77279-562 77279-564 77279-566
2-(1-Piperazinylcarbonyl)-1,4-benzodioxane hydrochloride 98%
2-(1-Piperazinylcarbonyl)-1,4-benzodioxane hydrochloride

Precaución: for Research Use Only

Formula: C₁₃H₁₆N₂O₃
MDL Number: MFCD00729051
CAS Number: 70918-74-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR