2-Methylcyclohexanone 95%

Supplier: Ambeed
Danger

A486727-1G A486727-25G A486727-5G A486727-100G
77279-556EA 11 USD
77279-556 77279-558 77279-560 77617-110
2-Methylcyclohexanone 95%
2-Methylcyclohexanone

Precaución: for Research Use Only

Formula: C₇H₁₂O
MDL Number: MFCD00001635
CAS Number: 583-60-8
UN: 1224
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR