2',5'-Difluoroacetanilide 97%

Supplier: Ambeed
Warning

Synonyms: N-(2,5-Difluorophenyl)acetamide

A486530-10G A486530-1G A486530-25G A486530-5G
77253-150EA 71.07 USD
77253-150 77253-152 77253-154 77253-156
2',5'-Difluoroacetanilide 97%
2',5'-Difluoroacetanilide

Precaución: for Research Use Only

Formula: C₈H₇F₂NO
MW: 171.1441 g/mol
MDL Number: MFCD00032512
CAS Number: 398-90-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR