4-Ethylbenzenesulfonamide 97%

Supplier: Ambeed
Warning

A486002-100G A486002-10G A486002-25G A486002-5G
77292-006EA 93.73 USD
77292-006 77292-008 77292-010 77292-012
4-Ethylbenzenesulfonamide 97%
4-Ethylbenzenesulfonamide

Precaución: for Research Use Only

Formula: C₈H₁₁NO₂S
MW: 185.24 g/mol
MDL Number: MFCD01365828
CAS Number: 138-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR