Ethyl (Z)-3-aminobut-2-enoate 98%

Supplier: Ambeed
Warning

A485752-1000G A485752-100G A485752-10G A485752-25G A485752-500G A485752-5G
77291-994EA 114.33 USD
77291-994 77291-996 77291-998 77292-000 77292-002 77292-004
Ethyl (Z)-3-aminobut-2-enoate 98%
Ethyl (Z)-3-aminobut-2-enoate

Precaución: for Research Use Only

Formula: C₆H₁₁NO₂
MW: 129.16 g/mol
MDL Number: MFCD02730138
CAS Number: 626-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR