Madrasin 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-((7-Methoxy-4-methylquinazolin-2-yl)amino)-5,6-dimethylpyrimidin-4(3H)-one
A484984-10MG
A484984-1MG
A484984-25MG
A484984-50MG
A484984-5MG
77241-360EA
92.7
USD
77241-360
77241-362
77241-364
77241-366
77241-368
Madrasin 98%
Madrasin
Precaución: for Research Use Only
Formula:
C₁₆H₁₇N₅O₂ MW: 311.34 g/mol |
MDL Number:
MFCD02233199 CAS Number: 374913-63-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...