5,5'-Bis(trimethylstannyl)-2,2'-bithiophene 97%

Supplier: Ambeed
Danger

A484785-250MG A484785-1G
77253-148EA 28 USD
77253-148 77551-470
5,5'-Bis(trimethylstannyl)-2,2'-bithiophene 97%
5,5'-Bis(trimethylstannyl)-2,2'-bithiophene

Precaución: for Research Use Only

Formula: C₁₄H₂₂S₂Sn₂
MDL Number: MFCD21363046
CAS Number: 143367-56-0
UN: 3146
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR