(±)-Indoline-2-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: Indoline-2-carboxylic acid

A484578-10G A484578-1G A484578-25G A484578-5G
77279-516EA 20.6 USD
77279-516 77279-518 77279-520 77279-522
(±)-Indoline-2-carboxylic acid 95%
(±)-Indoline-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₉NO₂
MW: 163.18 g/mol
MDL Number: MFCD00010635
CAS Number: 78348-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR