3,3'-Dimethyldiphenylamine 97%

Supplier: Ambeed
Warning

Synonyms: Di-m-tolylamine

A484491-1G A484491-250MG A484491-25G A484491-5G
77279-508EA 18.54 USD
77279-508 77279-510 77279-512 77279-514
3,3'-Dimethyldiphenylamine 97%
3,3'-Dimethyldiphenylamine

Precaución: for Research Use Only

Formula: C₁₄H₁₅N
MW: 197.28 g/mol
MDL Number: MFCD00059315
CAS Number: 626-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR