Chlorotris(triphenylphosphine)copper 97%

Supplier: Ambeed
Warning

A484449-100G A484449-1G A484449-5G A484449-25G
77265-532EA 593 USD
77265-532 77265-534 77265-536 77617-098
Chlorotris(triphenylphosphine)copper 97%
Chlorotris(triphenylphosphine)copper

Precaución: for Research Use Only

Formula: C₅₄H₄₅ClCuP₃
MW: 885.85 g/mol
MDL Number: MFCD00010270
CAS Number: 15709-76-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR