2-Chloro-4-methylquinazoline 97%

Supplier: Ambeed
Warning

A484154-100MG A484154-1G A484154-250MG
77265-526EA 43.26 USD
77265-526 77265-528 77265-530
2-Chloro-4-methylquinazoline 97%
2-Chloro-4-methylquinazoline

Precaución: for Research Use Only

Formula: C₉H₇ClN₂
MW: 178.61 g/mol
MDL Number: MFCD09998826
CAS Number: 6141-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR