(E)-3-((tert-Butoxycarbonyl)(isopropyl)amino)-2-(4-chlorophenyl)acrylic acid 97%

Supplier: Ambeed
Warning

A484099-1G A484099-250MG
77265-522EA 21.63 USD
77265-522 77265-524
(E)-3-((tert-Butoxycarbonyl)(isopropyl)amino)-2-(4-chlorophenyl)acrylic acid 97%
(E)-3-((tert-Butoxycarbonyl)(isopropyl)amino)-2-(4-chlorophenyl)acrylic acid

Precaución: for Research Use Only

Formula: C₁₇H₂₂ClNO₄
MW: 339.81 g/mol
MDL Number: MFCD29078977
CAS Number: 1489004-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR