2-Chlorobenzenesulfonyl chloride 96%

Supplier: Ambeed
Danger

A483968-10G A483968-1G A483968-25G A483968-5G
77291-962EA 23 USD
77291-962 77291-964 77291-966 77291-968
2-Chlorobenzenesulfonyl chloride 96%
2-Chlorobenzenesulphonyl chloride

Precaución: for Research Use Only

Formula: C₆H₄Cl₂O₂S
MDL Number: MFCD00051520
CAS Number: 2905-23-9
UN: 2920
ADR: 8,3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR