2-(Bromomethyl)-5-methylpyridine hydrobromide 97%

Supplier: Ambeed
Danger

A483839-100MG A483839-1G
77265-510EA 99 USD
77265-510 77594-506
2-(Bromomethyl)-5-methylpyridine hydrobromide 97%
2-(Bromomethyl)-5-methylpyridine hydrobromide

Precaución: for Research Use Only

Formula: C₇H₉Br₂N
CAS Number: 2089315-69-3
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR